Density Sets
Density Set, Polypropylene (PP)
470221-110
Supplier #: PH0113H
Students Learn That the Density of a Substance is Constant, Even as Its Mass and Volume Change.
- Each set has 12 cylindrical bars
- Bars are same diameter but different lengths
- Each set comes in an attractive wooden holder
- Description:Density Set, Polypropylene (PP)