Order Entry
Mexico
Orders LinkContactUsLinkComponent
Clean Scrub™ Mop, Contec®

Clean Scrub™ Mop, Contec®

Clean Scrub™ Sponge, 13 x 8 x 1.5 cm (5 x 3 x 0.6")
Catalog # 10811-978
Supplier:   CONTEC, INC. MX
Supplier #:  VKSC0004
Clean Scrub™ Mop, Contec®
Catalog # 10811-978
Supplier:  CONTEC, INC. MX
Clean Scrub™ Mop, Contec®
Clean Scrub™ Mop, Contec®

Clean Scrub™ Sponge, 13 x 8 x 1.5 cm (5 x 3 x 0.6")

Price
$6,196.84
Log in for availability
Case (48UN)
Product Details & Documents

Contec’s Clean Scrub™ single-use mop head is wrapped with a textured polyester mesh to eliminate dirt and grime from heavily contaminated surfaces. The mop head has a core of polyester fabric and foam that provides exceptional liquid absorption and resists tearing.

  • Textured polyester mesh removes heavily soiled contamination from surfaces
  • Single-use mop head eliminates cross contamination
  • Slim profile fits into small spaces
  • Compatible with a wide range of disinfectants

This pad is fastened to a semi-rigid thermoformed plate that easily snaps onto existing QuickConnect mop frames and provides steady contact pressure on mopped surfaces. The Clean Scrub™ mop will not scratch surfaces. Compatible with most disinfectants and solutions, Clean Scrub™ is ideal for many cleaning applications. Due to the thermoformed plastic backplate, Clean Scrub™ should not be autoclaved.

Specifications
  • Description:
    Clean Scrub™ Sponge
  • Size:
    13 x 8 x 1.5 cm (5 x 3 x 0.6")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.