Optical Refraction Components
Double Concave, Acrylic, frosted, –200 mm Focal Length
470219-496
: PH0577B
These Optical Refraction Components are made from imported optical glass or acrylic with optically worked surfaces.
- Students Measure the Refraction of Light
- Refraction Blocks, Prisms, and Lenses
- Optically Worked Surfaces
- Bubble-Free Glass
- Ideal in Conjunction with a Light Box
All angles are accurately worked with edges slightly beveled and glass free from bubbles.
These optical elements are perfect to use with a light box, so students can study and measure the refraction of light through different mediums and different shapes.
- Description:Double Concave
- Dimensions:–200 mm Focal Length
- Material:Acrylic, frosted