Order Entry
Mexico
Orders LinkContactUsLinkComponent
Optical Refraction Components
Optical Refraction Components
Double Convex, Acrylic, frosted, 200 mm Focal Length
Catalog # 470219-494
Supplier #:  PH0577A
Optical Refraction Components
Catalog # 470219-494
Optical Refraction Components
Optical Refraction Components
Double Convex, Acrylic, frosted, 200 mm Focal Length
Price
$411.48
Log in for availability
Each (1UN)
Product Details & Documents
These Optical Refraction Components are made from imported optical glass or acrylic with optically worked surfaces.
  • Students Measure the Refraction of Light
  • Refraction Blocks, Prisms, and Lenses
  • Optically Worked Surfaces
  • Bubble-Free Glass
  • Ideal in Conjunction with a Light Box
All angles are accurately worked with edges slightly beveled and glass free from bubbles.
These optical elements are perfect to use with a light box, so students can study and measure the refraction of light through different mediums and different shapes.
Specifications
  • Description:
    Double Convex
  • Dimensions:
    200 mm Focal Length
  • Material:
    Acrylic, frosted