Optical Prisms
Glass, 50 mm
Catalog # 470219-450
Supplier: EISCO SCIENTIFIC LLC
Supplier #: PH0558C
Jump to:
Product Details & Documents
Right angle and equilateral prisms are available in a variety of forms.
- For Investigating Refraction and Creating Rainbows
- Non-Optically Worked
- Polished Faces
- Beveled Edges
- Right Angle and Equilateral Prisms Available
These prisms are non-optically worked with polished faces and edges slightly beveled. They're useful for investigating refraction and creating rainbows.
Choose the size and material that best fits your experimental needs.
Specifications
- Material:Glass
- Size:50 mm