General Purpose Viscosity Standards, Cannon Instruments
Product Details & Documents
These certified standards are useful for calibration and verification procedures.
- It is suitable for all types of viscometers
- It also helps ensures the reliability of measurements
Certifications: Meets the precision specifications of ASTM D 445/446 and ISO 3104/3105, and are traceable to the NIST.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Description:Viscosity standard S60
- Size:500 ml
- Viscosity:(20°C) 170 mPa·s