Glassware Washers Accessories
Basket cover, stainless steel, ½ size baskets (10002-908, 10002-910, 10002-912, 10002-924)
Catalog # 10002-944
Supplier: VWR International
Jump to:
Product Details & Documents
Accessories designed for use with VWR® Undercounter, Freestanding and Vertical Stacking Glassware washers safeguard flasks, beakers, test tubes, bottles, Petri dishes, and more.
- Accessories maximize space, improve efficiency, and protect specialty glassware; use in standard open racks
- Bottom mesh inserts prevent small items from falling through
- Made of stainless steel for long life and durability
- Booster pump for use with low pressure DI water supply below 18 psi
See details in table below for specific use information. For further assistance, view the product literature or contact your Avantor representative.
Specifications
- Description:Basket cover, stainless steel
- Fits:½ size baskets (10002-908, 10002-910, 10002-912, 10002-924)