Dropping Bottles
Amber, 30 ml
Catalog # 470217-652
Supplier: EISCO SCIENTIFIC LLC
Supplier #: CH0187E
Jump to:
Product Details & Documents
Dropping bottles are available in clear or amber formats.
- Screw cap included
- Dropper included
Bottles are made of soda glass.
Specifications
- Description:Amber, 30 ml