Order Entry
Mexico
Orders LinkContactUsLinkComponent
Tripod Triangle Stands

Tripod Triangle Stands

Tripod Triangle Stand for burners, 21 cm, 12 cm
Catalog # 470165-364
Supplier #:  CH0645A
Tripod Triangle Stands
Catalog # 470165-364
Tripod Triangle Stands
Tripod Triangle Stands

Tripod Triangle Stand for burners, 21 cm, 12 cm

Price
$233.46
Log in for availability
Each (1UN)
Product Details & Documents

Zinc plated stands feature cast iron top and mild steel legs. Multiple sizes available.

Specifications
  • Ext.Ø:
    12 cm
  • Description:
    Tripod Triangle Stand for burners
  • Height:
    21 cm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.