Order Entry
Mexico
Orders LinkContactUsLinkComponent
Nylon Burette Brushes

Nylon Burette Brushes

Nylon burette brush, for 50 mL burettes, Nylon, 19 mm, Stainless steel, 760 mm
Catalog # 470165-360
Supplier #:  CH0217E
Nylon Burette Brushes
Catalog # 470165-360
Nylon Burette Brushes
Nylon Burette Brushes

Nylon burette brush, for 50 mL burettes, Nylon, 19 mm, Stainless steel, 760 mm

Price
$101.26
Log in for availability
Each (1UN)
Product Details & Documents

These brushes are available in a variety of sizes.

  • Durable
  • Long-lasting nylon bristles

These durable brushes will keep your burettes and labware clean.

Specifications
  • Bristle Material:
    Nylon
  • Brush head Ø:
    19 mm
  • Handle Material:
    Stainless steel
  • Description:
    Nylon burette brush, for 50 mL burettes
  • Total length:
    760 mm