Liquid Sampler, Bürkle
100 ml, 53.5 cm
10217-128
Samplers for thin to viscous liquids.
- Liquid Sampler, open with thumb: The valve is opened with the thumb to empty the sample
- Liquid Sampler, place onto bottle: The valve is opened by attaching the sample container to empty the sample
Easy use with one-handed operation. With discharge funnel for safe filling of the sample bottles. Material V4A (1.4404) ; Container Diameter: 32 mm
- Capacity:100 ml
- Length:53.5 cm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



