Order Entry
Mexico
Orders LinkContactUsLinkComponent
Eisco® Human Gametes Models

Eisco® Human Gametes Models

Egg, 1, 13 cm, 13 cm, 18 cm
Catalog # 470136-174
Supplier #:  AM0115B
Eisco® Human Gametes Models
Catalog # 470136-174
Eisco® Human Gametes Models
Eisco® Human Gametes Models

Egg, 1, 13 cm, 13 cm, 18 cm

Price
$1,982.25
Log in for availability
Each (1UN)
Product Details & Documents

Human egg and sperm gametes with cutaway views for internal study.

  • Cutaway sections reveal internal structures
  • Once piece models are rod mounted on a base
  • Designed by medical professionals and educators
  • Custom molded using the highest quality nontoxic polymers and PVC
  • Hand-painted by skilled artists
  • Each model includes a key card

Greatly magnified to show structures, these models features a cut away section to show internal features. Choose the sperm, the ovum, or both to help illustrate human reproductive cells.

Specifications
  • Description:
    Egg
  • Height:
    18 cm
  • Length:
    13 cm
  • Width:
    13 cm
  • No. of Parts:
    1