Order Entry
Mexico
Orders LinkContactUsLinkComponent
Dropping Bottles

Dropping Bottles

30 ml (1 oz.), 18-400
Catalog # 470123-899
Supplier:   QORPAK
Supplier #:  501181
Dropping Bottles
Catalog # 470123-899
Supplier:  QORPAK
Dropping Bottles
Dropping Bottles

30 ml (1 oz.), 18-400

Price
$1,162.51
Log in for availability
Case (24UN)
Product Details & Documents

Dropping bottles are made of low-density polyethylene.

  • White Polypropylene Dropper and Nasal Cap
  • Molded Plastic Screw Cap Seals at Tip
  • Low-Density Polyethylene

They feature a separable dropping tip.

The molded plastic screw cap seals at the tip.

Documents

Specifications
  • Cap Size:
    18-400
  • Capacity:
    30 ml (1 oz.)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.