Order Entry
Mexico
Orders LinkContactUsLinkComponent
Dropper Bottles
Dropper Bottles
Dropper Bottles, 8 ml
Catalog # 470211-796
Supplier #:  2752-9025
Dropper Bottles
Catalog # 470211-796
Dropper Bottles
Dropper Bottles
Dropper Bottles, 8 ml
Price
$1,790.00
Log in for availability
Case (25UN)
Product Details & Documents
These bottles are made of low-density polyethylene.
  • Bottles Dispense about 40 mcL per Drop
  • Good Chemical Resistance
  • Semi-Transparent Bottles
  • Polyethylene Construction
The semi-transparent bottles feature color-coded caps.
They have good chemical resistance and dispense about 40 mcL per drop.
Specifications
  • Capacity:
    8 ml
  • Description:
    Dropper Bottles