Order Entry
Mexico
Orders LinkContactUsLinkComponent
PYREX® Watch Glasses
PYREX® Watch Glasses
Watch glass, 65 mm
Catalog # 470211-706
Supplier:   CORNING CA
Supplier #:  9985-65
PYREX® Watch Glasses
Catalog # 470211-706
Supplier:  CORNING CA
PYREX® Watch Glasses
PYREX® Watch Glasses
Watch glass, 65 mm
Price
$1,330.00
Log in for availability
Each (12UN) Paquete de 12 Case de 12 (12UN)
Product Details & Documents
PYREX® borosilicate glass.
  • High chemical durability
  • Prevents contamination of solutions
These watch glasses are molded to the same radius of curvature for convenient stacking. The heavy wall and uniform fire-polished edge provides the mechanical strength. The outside diameter cannot be controlled within close limits and a slight degree of out-of-round can be expected.
They are also used as covers for heated beakers.
Packaging: One package has 12 watch glasses.
Documents
Specifications
  • Ø:
    65 mm
  • Description:
    Watch glass