Order Entry
Mexico
Orders LinkContactUsLinkComponent
PYREX® Watch Glasses

PYREX® Watch Glasses

Watch glass, 100 mm
Catalog # 470211-702
Supplier #:  9985-100
PYREX® Watch Glasses
Catalog # 470211-702
Supplier:  
PYREX® Watch Glasses
PYREX® Watch Glasses

Watch glass, 100 mm

Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
WATCH GLASS 100MM 9985 12/PK Catalog #66110-087

Product 66110-087 is similar to 470211-702 but may have a different manufacturer and/or packaging.

Product Details & Documents

PYREX® borosilicate glass.

  • High chemical durability
  • Prevents contamination of solutions

These watch glasses are molded to the same radius of curvature for convenient stacking. The heavy wall and uniform fire-polished edge provides the mechanical strength. The outside diameter cannot be controlled within close limits and a slight degree of out-of-round can be expected.

They are also used as covers for heated beakers.

 : One package has 12 watch glasses.

Documents

Specifications
  • Ø:
    100 mm
  • Description:
    Watch glass