PYREX® Volumetric Pipettes, Color-Coded, Class A
Pyrex® transfer pipettes (volumetric pipettes), cap., each, 1 ml
Supplier #: 7100-1
PYREX® Volumetric Pipettes, Color-Coded, Class A
Catalog # 470211-598
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
PIPETTE BOROSILICATE CLASS A BLUE 1 ML Catalog #470312-906
Product 470312-906 is a replacement for 470211-598 and has the same packaging and specifications, but may be a different quantity.
VWR PIPET VOLMTRC TRANSFR 1ML CS12 Catalog #53044-037
Product 53044-037 is a replacement for 470211-598 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
These pipettes are made of high-grade borosilicate glass.
- Reusable, rugged construction
- Color-coded
- Colored enamel graduations and markings
Ruggedly constructed and properly proportioned, pipettes have accuracy tolerances to meet or exceed the strict requirements for Class A Volumetric Ware as indicated by ASTM E969.
Documents
Certificates
Search by Lot NumberSpecifications
- Capacity:1 ml
- Description:Pyrex® transfer pipettes (volumetric pipettes), cap., each
- Cat. No.:470211-598
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Environmentally Preferable:Low Manufacturing Impact