Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

PYREX® Volumetric Flasks

2000 ml
Catalog # 470211-516
   Low Manufacturing Impact
Supplier:   CORNING CA
 :  5580-2L
PYREX® Volumetric Flasks
Catalog # 470211-516
Supplier:  CORNING CA
   Low Manufacturing Impact
PYREX® Volumetric Flasks
PYREX® Volumetric Flasks

2000 ml

Price
$3,020.00
Log in for availability
Each (1UN) Case de 4 (1UN)
Product Details & Documents

PYREX® glass, clear, with snap-caps.

  • Heavy beads and tubing rings tooled for snap-cap fit
  • Graduation line is sharp and permanent
  • No snap caps for 1 and 2 ml sizes
Specifications
  • Capacity:
    2000 ml
  • Cat. No.:
    470211-516
  • Environmental certification:
    I-RECs, ISO 14001 - Monterrey
  • Environmentally Preferable:
    Low Manufacturing Impact