PYREX® Volumetric Flasks
100 ml
Supplier: CORNING CA
Supplier #: 5580-100
Jump to:
Product Details & Documents
PYREX® glass, clear, with snap-caps.
- Heavy beads and tubing rings tooled for snap-cap fit
- Graduation line is sharp and permanent
- No snap caps for 1 and 2 ml sizes
Documents
Specifications
- Capacity:100 ml
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



