Order Entry
Mexico
Orders LinkContactUsLinkComponent
Pyrex® Erlenmeyer Flasks

Pyrex® Erlenmeyer Flasks

500 ml, 50 ml, 100 - 500 ml, 7
Catalog # 470211-492
Supplier:   CORNING CA
Supplier #:  4980-500
Pyrex® Erlenmeyer Flasks
Catalog # 470211-492
Supplier:  CORNING CA
Pyrex® Erlenmeyer Flasks
Pyrex® Erlenmeyer Flasks

500 ml, 50 ml, 100 - 500 ml, 7

Price
$1,420.00
Log in for availability
Paquete de 6 Case de 6 (6UN)
Product Details & Documents

PYREX® borosilicate glass 3.3.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

Heavy duty rim flasks have approximate graduations in durable white enamel. Extra large marking space. Heavy, durable top tooling.

Specifications
  • Graduation Interval:
    50 ml
  • Graduation Range:
    100 - 500 ml
  • Capacity:
    500 ml
  • Stopper No.:
    7
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.