Order Entry
Mexico
Orders LinkContactUsLinkComponent
Pyrex® Erlenmeyer Flasks

Pyrex® Erlenmeyer Flasks

50 ml, 10 ml, 20 - 50 ml, 1
Catalog # 470211-490
Supplier:   CORNING CA
Supplier #:  4980-50
Pyrex® Erlenmeyer Flasks
Catalog # 470211-490
Supplier:  CORNING CA
Pyrex® Erlenmeyer Flasks
Pyrex® Erlenmeyer Flasks

50 ml, 10 ml, 20 - 50 ml, 1

Price
$2,510.00
Log in for availability
Paquete de 12 Case de 4 (12UN)
Product Details & Documents

PYREX® borosilicate glass 3.3.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

Heavy duty rim flasks have approximate graduations in durable white enamel. Extra large marking space. Heavy, durable top tooling.

Specifications
  • Graduation Interval:
    10 ml
  • Graduation Range:
    20 - 50 ml
  • Capacity:
    50 ml
  • Stopper No.:
    1
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.