Order Entry
Mexico
Orders LinkContactUsLinkComponent
Pyrex® Erlenmeyer Flasks

Pyrex® Erlenmeyer Flasks

25 ml, 5 ml, 10 - 25 ml, 0
Catalog # 470211-484
Supplier:   CORNING CA
Supplier #:  4980-25
Pyrex® Erlenmeyer Flasks
Catalog # 470211-484
Supplier:  CORNING CA
Pyrex® Erlenmeyer Flasks
Pyrex® Erlenmeyer Flasks

25 ml, 5 ml, 10 - 25 ml, 0

Price
$2,570.00
Log in for availability
Paquete de 12 Case de 4 (12UN)
Product Details & Documents

PYREX® borosilicate glass 3.3.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

Heavy duty rim flasks have approximate graduations in durable white enamel. Extra large marking space. Heavy, durable top tooling.

Specifications
  • Graduation Interval:
    5 ml
  • Graduation Range:
    10 - 25 ml
  • Capacity:
    25 ml
  • Stopper No.:
    0