Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Pyrex® Erlenmeyer Flasks

125 ml, 25 ml, 50 - 125 ml, 5
  470211-480
 :   CORNING CA
 :  4980-125
Pyrex® Erlenmeyer Flasks
  470211-480
Pyrex® Erlenmeyer Flasks
Pyrex® Erlenmeyer Flasks

125 ml, 25 ml, 50 - 125 ml, 5

 
$2,370.00
   
Paquete de 12 Case de 4 (12UN)
 

PYREX® borosilicate glass 3.3.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

Heavy duty rim flasks have approximate graduations in durable white enamel. Extra large marking space. Heavy, durable top tooling.

 
  • Graduation Interval:
    25 ml
  • Graduation Range:
    50 - 125 ml
  • Capacity:
    125 ml
  • Stopper No.:
    5
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.