Order Entry
Mexico
Orders LinkContactUsLinkComponent
Bottles, Square, Polycarbonate, Corning

Bottles, Square, Polycarbonate, Corning

Corning Square Polycarbonate Bottles, 500 mL
Catalog # 470211-302
Supplier:   CORNING CA
Supplier #:  431432
Bottles, Square, Polycarbonate, Corning
Catalog # 470211-302
Supplier:  CORNING CA
Bottles, Square, Polycarbonate, Corning
Bottles, Square, Polycarbonate, Corning

Corning Square Polycarbonate Bottles, 500 mL

Price
$211.00
Log in for availability
Each (1UN) Case de 24 (1UN)
Product Details & Documents

PC, square.

  • More break-resistant than other glass or plastic bottles
  • Screened white enamel graduations are easier to see than moulded graduations
  • Large white marking spot for easier identification
  • Bottles can be autoclaved once at 121 °C and 15 psi (repeated autoclaving weakens PC and is not recommended)
  • Can be stored at –80 °C

These storage bottles are easier to handle, require less space on the shelf or in the autoclave, and are ideal for mixing, sampling and storage.

Documents

Specifications
  • Description:
    Corning Square Polycarbonate Bottles, 500 mL