Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

LeLoop® Inoculating Loops

Non-Calibrated Loop, Large, 5.45 µl (3.57 mm)
  89426-286
 :   DECON LABS
 :  MP19825
LeLoop® Inoculating Loops
  89426-286
LeLoop® Inoculating Loops
LeLoop® Inoculating Loops

Non-Calibrated Loop, Large, 5.45 µl (3.57 mm)

 
$2,560.00
   
Paquete de 25
 
   
 

LeLoop®, Decon’s inoculating loops, are made of high-quality, twisted nichrome wire which is designed to cool rapidly after flaming.

  • Small loop: 2.26 mm Int.Ø

  • Medium loop: 2.91 mm IInt.Ø
  • Large loop: 3.57 mm Int.Ø

These loops are more economical than platinum or disposable plastic loops. The loops slide smoothly across the agar surface without tearing or gouging. Decon loops are made to be used with Decon’s brass holders.

Calibrated 1 µl (0.001 ml) and 10 µl (0.01 ml) inoculating loops are individually calibrated in accordance with ASM/APHA via Evans Blue Dye Test which is a CAP approved method. Calibration certificates are sent with calibrated loops or are available from Decon Labs.

Certifications: UL listed.

Documents

 
 
 
  • Capacity:
    5.45 µl (3.57 mm)
  • Description:
    Non-Calibrated Loop, Large
Accessories

 

  •  
    Description
     
     
  • 89426-274
    6" Brass Holder
       
    Each (1UN) - $1,280.00

 

  •  
    Description
     
     
  • 89426-276
    8" Brass Holder
       
    Each (1UN) - $1,820.00

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.