Order Entry
Mexico
Orders LinkContactUsLinkComponent
LeLoop® Inoculating Loops

LeLoop® Inoculating Loops

Non-Calibrated Loop, Small, 2.45 µl (2.26 mm)
Catalog # 89426-282
Supplier:   DECON LABS
Supplier #:  MP19725
LeLoop® Inoculating Loops
Catalog # 89426-282
Supplier:  DECON LABS
LeLoop® Inoculating Loops
LeLoop® Inoculating Loops

Non-Calibrated Loop, Small, 2.45 µl (2.26 mm)

Price
$2,560.00
Log in for availability
Paquete de 25
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

LeLoop®, Decon’s inoculating loops, are made of high-quality, twisted nichrome wire which is designed to cool rapidly after flaming.

  • Small loop: 2.26 mm Int.Ø

  • Medium loop: 2.91 mm IInt.Ø
  • Large loop: 3.57 mm Int.Ø

These loops are more economical than platinum or disposable plastic loops. The loops slide smoothly across the agar surface without tearing or gouging. Decon loops are made to be used with Decon’s brass holders.

Calibrated 1 µl (0.001 ml) and 10 µl (0.01 ml) inoculating loops are individually calibrated in accordance with ASM/APHA via Evans Blue Dye Test which is a CAP approved method. Calibration certificates are sent with calibrated loops or are available from Decon Labs.

Certifications: UL listed.

Documents

Certificates
Search by Lot Number
Specifications
  • Capacity:
    2.45 µl (2.26 mm)
  • Description:
    Non-Calibrated Loop, Small
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • 6" Brass Holder
    6" Brass Holder
    89426-274
    Log in for availability
    Each (1UN) - $1,280.00

Product Information

  • Catalog #
    Availability
    Price
  • 8" Brass Holder
    8" Brass Holder
    89426-276
    Log in for availability
    Each (1UN) - $1,820.00

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.