Order Entry
Mexico
Orders LinkContactUsLinkComponent
Media Bottles
Media Bottles
500 ml, Media Bottle
Catalog # 470205-568
Supplier #:  219759
Media Bottles
Catalog # 470205-568
Media Bottles
Media Bottles
500 ml, Media Bottle
Price
$370.29
Log in for availability
Each (1UN) Case de 24 (1UN)
Product Details & Documents
Borosilicate Glass.
  • Large opening, no-drip-lip, and sloping shoulders for easy pouring
  • Approximate volume graduations
  • Large writing area
Both the bottles and rubber-lined caps are autoclavable.
Specifications
  • Capacity:
    500 ml
  • Description:
    Media Bottle