Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Economical Class 7 Hooked Weights

Hooked Weights, 10 g to 1000 g, 9 weights
  470204-266
 :  30390775
Economical Class 7 Hooked Weights
  470204-266
Economical Class 7 Hooked Weights
Economical Class 7 Hooked Weights

Hooked Weights, 10 g to 1000 g, 9 weights

 
$6,316.16
   
Kit de 1
 

For rough weighing operations in physical and chemical laboratories, such as force measuring apparatus.

  • Weights above 5 g are constructed of 303 Stainless Steel
  • Weights 5 g and below are constructed of 3003-H14 Aluminum
  • Manufactured in the USA

All weights are hooked and feature two-piece construction. Weight sets are supplied in a weight block with compartments for each weight. Individual weights are not supplied in a case.

 
  • Description:
    Hooked Weights, 10 g to 1000 g
  • No. of Weights:
    9 weights
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.