Conductivity/TDS Calibration Solution
Conductivity/TDS Calibration Solution, 1413 μS, 989 ppm
Catalog # 470203-116
Supplier: LA MOTTE CHEMICAL PROD CO SE
Supplier #: 6354-L
Conductivity/TDS Calibration Solution
Catalog # 470203-116
Supplier: LA MOTTE CHEMICAL PROD CO SE
CAS Number:
Jump to:
Product Details & Documents
Always keep your meter in range.
- Already prepared solutions
- Various ranges
- Aligned with GLOBE protocols
Prepared solution to keep your conductivity calibrated. 500ml bottles.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Description:Conductivity/TDS Calibration Solution
- Conductivity:1413 μS
- Dissolved Solids:989 ppm



