Order Entry
Mexico
Orders LinkContactUsLinkComponent
Nylon Beaker Brushes

Nylon Beaker Brushes

Nylon Beaker Brush, Heavy Double Tufted End, Nylon, Black, 12.7 cm, 76.2 mm (3”), Wood, 40 cm
Catalog # 470202-910
Supplier #:  J720400
Nylon Beaker Brushes
Catalog # 470202-910
Nylon Beaker Brushes

Nylon Beaker Brush, Heavy Double Tufted End, Nylon, Black, 12.7 cm, 76.2 mm (3”), Wood, 40 cm

Price
$664.80
Log in for availability
Each (1UN)
Product Details & Documents

For beakers, jars, cylinders, etc. With black nylon fibers and stiff tufted end on a wood handle.

Specifications
  • Bristle Color:
    Black
  • Bristle Material:
    Nylon
  • Brush head Ø:
    76.2 mm (3”)
  • Handle Material:
    Wood
  • Description:
    Nylon Beaker Brush, Heavy Double Tufted End
  • Total length:
    40 cm
  • Brush Head Length:
    12.7 cm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.