PYREX® Erlenmeyer Flask with Screw Cap
500 ml
Supplier: CORNING CA
Supplier #: 4985-500
Jump to:
Product Details & Documents
Made of PYREX® borosilicate glass, with phenolic screw caps.
- Excellent chemical resistance
- Excellent temperature resistance
- Graduated to show approximate capacity
- With marking spot
Erlenmeyer flasks with phenolic screw caps are ideal for use as a shaker flask or for mixing, media prep and storage needs. Impact resistant caps are equipped with cemented-in, inert white rubber liners which are resistant to steam sterilization. Their uniform wall thickness provides the proper balance between mechanical strength and thermal shock resistance.
Documents
Certificates
Search by Lot NumberSpecifications
- Capacity:500 ml