Order Entry
Mexico
Orders LinkContactUsLinkComponent
PYREX® Erlenmeyer Flask with Screw Cap
PYREX® Erlenmeyer Flask with Screw Cap
1000 ml
Catalog # 470202-216
Supplier:   CORNING CA
Supplier #:  4985-1L
PYREX® Erlenmeyer Flask with Screw Cap
Catalog # 470202-216
Supplier:  CORNING CA
PYREX® Erlenmeyer Flask with Screw Cap
PYREX® Erlenmeyer Flask with Screw Cap
1000 ml
Price
$1,240.00
Log in for availability
Each (1UN) Paquete de 6 (1UN) Case de 12 (1UN)
Product Details & Documents
Made of PYREX® borosilicate glass, with phenolic screw caps.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Graduated to show approximate capacity
  • With marking spot
Erlenmeyer flasks with phenolic screw caps are ideal for use as a shaker flask or for mixing, media prep and storage needs. Impact resistant caps are equipped with cemented-in, inert white rubber liners which are resistant to steam sterilization. Their uniform wall thickness provides the proper balance between mechanical strength and thermal shock resistance.
Documents
Specifications
  • Capacity:
    1000 ml