Order Entry
Mexico
Orders LinkContactUsLinkComponent
Flask Brushes
Flask Brushes
White, Plastic, 17"
Catalog # 470201-504
Supplier:   ALLIED K & R
Supplier #:  D132
Flask Brushes
Catalog # 470201-504
Supplier:  ALLIED K & R
Flask Brushes
Flask Brushes
White, Plastic, 17"
Price
$148.24
Log in for availability
Each (1UN) Case de 144 (1UN)
Product Details & Documents
White bristle and plastic handle.
  • Multiple sizes available
Recommended for volumetric and boiling flasks because of the unusally flexible, curved steel spring that enables the brush to reach all parts of the flask.
Specifications
  • Bristle Color:
    White
  • Handle Material:
    Plastic
  • Total length:
    17"