Polymethylpentene Cylinders
100 ml, 250 mm, 31 mm, 1 ml, 1 ml
470199-962
: DYNALON
: 237765
Glass-Like Clarity.
- Permanently molded graduations
- Autoclavable
The cylinders are chemically resistant to most acids, bases, alcohols, and aldehydes.
- Ø:31 mm
- Graduation Range:1 ml
- Capacity:100 ml
- Height:250 mm
- Tolerance:1 ml



