Clostridium botulinum
Clostridium botulinum, Smear, Gram (+)
Catalog # 470199-728
Supplier: Avantor
Jump to:
Product Details & Documents
Potentially pathogenic bacteria
- Identifying features clearly distinguishable
- Rigorous quality control standards
Rods. Endospores oval and subterminal. Agent of botulism.
Specifications
- Description:Clostridium botulinum
- Stain:Gram (+)
- Orientation:Smear