Order Entry
Mexico
Orders LinkContactUsLinkComponent
Polyethylene Wash Bottles
Polyethylene Wash Bottles
500 ml
Catalog # 470191-300
Supplier:   VWR International
Polyethylene Wash Bottles
Catalog # 470191-300
Supplier:  VWR International
Polyethylene Wash Bottles
Polyethylene Wash Bottles
500 ml
Price
$39.22
Log in for availability
Each (1UN) Case de 24 (1UN)
Product Details & Documents
Unbreakable wash bottles for student use.
  • Polyethylene, narrow mouth construction
  • Closures included
Cap and delivery tube are one-piece construction to eliminate leakage. The inside diameter of the delivery tip is carefully drawn to give a fine stream of liquid.
Documents
Specifications
  • Capacity:
    500 ml