Order Entry
Mexico
Orders LinkContactUsLinkComponent
VWR® Volumetric Flasks, Class B
VWR® Volumetric Flasks — Class B, Cap., mL: 50, Tolerance, ±mL: 0.10
  470191-248
VWR® Volumetric Flasks, Class B
  470191-248
 :  
VWR® Volumetric Flasks, Class B
VWR® Volumetric Flasks — Class B, Cap., mL: 50, Tolerance, ±mL: 0.10
Our new high-quality borosilicate volumetric flasks meet ASTM standards and are a great value.
  • Borosilicate glass construction
  • Snap-on caps
  • Graduated line for accurate measurements
Class B with capacity tolerances twice those specified for Class A volumetric work. The necks are tooled to fit the included snap-caps. With graduation line for accurate measurements.
  • Description:
    VWR® Volumetric Flasks — Class B, Cap., mL: 50, Tolerance, ±mL: 0.10