Lens Paper
Lens Paper, Form: Booklet, Sheets, 6", 4"
Catalog # 470149-424
Supplier: RAYLABCON INC SE
: 20 20 5118
Jump to:
Product Details & Documents
This soft, absorbent paper cleans lenses and optical elements.
- Available in booklet and sheet form
The paper removes grit, lint, and dirt from cleaning lenses and optical elements.
Specifications
- Width:4"
- Description:Lens Paper, Form: Booklet, Sheets
- Length:6"
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



