Order Entry
Mexico
Orders LinkContactUsLinkComponent
Balls for Physics Experiments

Balls for Physics Experiments

Drilled Brass Ball, 25 mm, 1"
Catalog # 470007-150
Supplier #:  BLDBL25
Balls for Physics Experiments
Catalog # 470007-150
Balls for Physics Experiments
Balls for Physics Experiments

Drilled Brass Ball, 25 mm, 1"

Price
$247.84
Log in for availability
Each (1UN)
Product Details & Documents

Ideal for Collision and Pendulum Demonstrations

Drilled balls for pendulum bobs or impact experiments. 

Specifications
  • Description:
    Drilled Brass Ball, 25 mm
  • Diameter:
    1"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.