Order Entry
Mexico
Orders LinkContactUsLinkComponent
Ward's® Fluorite (Cleavage)
Fluorite (Cleavage), Hand Specimen, 2×3" - 3×4"
Catalog # 470025-580
Supplier:   Avantor
Ward's® Fluorite (Cleavage)
Catalog # 470025-580
Supplier:  Avantor
Ward's® Fluorite (Cleavage)
Ward's® Fluorite (Cleavage)
Fluorite (Cleavage), Hand Specimen, 2×3" - 3×4"
Price
$561.28
Log in for availability
Each (1UN)
Product Details & Documents
Pure, large, transparent-to-translucent cleavages, Mohs’ hardness of 4.
  • Collected in Mexico
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Specifications
  • Description:
    Fluorite (Cleavage), Hand Specimen
  • Specimen Dimensions:
    2×3" - 3×4"