Order Entry
Mexico
Orders LinkContactUsLinkComponent
Plastic Beads
Plastic Beads
Plastic Beads, 12 mm
Catalog # 470023-958
Supplier:   Avantor
Plastic Beads
Catalog # 470023-958
Supplier:  Avantor
Plastic Beads
Plastic Beads
Plastic Beads, 12 mm
Price
$487.98
Log in for availability
Paquete de 100
Product Details & Documents
Packages of approximately 100.
  • 4 mm Red bead = Acrylic
  • 8 mm Navy bead = Acrylic
  • 12 mm White bead = Acetate
  • Autoclavable
Specifications
  • Ø:
    12 mm
  • Description:
    Plastic Beads