Refraction Blocks
Refraction Block, 100 x 75 x 20 mm
Catalog # 470105-728
Supplier: UNITED SCIENTIFIC SUPPLIES
Supplier #: RCB100
Jump to:
Product Details & Documents
Rectangular blocks in a variety of sizes and are designed for use as an index of refraction plate.
- Quality Lucite block
- Optically clear sides
This Lucite block allows you to investigate refraction and internal reflection.
Specifications
- Description:Refraction Block
- Dimensions:100 x 75 x 20 mm