Order Entry
Mexico
Orders LinkContactUsLinkComponent
Ward's® Limestone (Encrinal)
Limestone (Encrinal), Student Specimens, 1×1 - 1×1"
  470026-086
 :   Avantor
Ward's® Limestone (Encrinal)
  470026-086
 :  Avantor
Ward's® Limestone (Encrinal)
Ward's® Limestone (Encrinal)
Limestone (Encrinal), Student Specimens, 1×1 - 1×1"
$267.48
Paquete de 10
Gray with line crinoid columnal fragments.
  • Collected in Ohio
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack - Includes 1 kg of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range. Some crinoids may require a hand lens to observe.
  • Description:
    Limestone (Encrinal), Student Specimens
  • Specimen Dimensions:
    1×1 - 1×1"