Order Entry
Mexico
Orders LinkContactUsLinkComponent
Ward's® Granite (Gray)
Ward's® Granite (Gray)
Granite (Gray), Bulk Pack, 1 kg, Size will vary
Catalog # 470026-012
Supplier:   Avantor
Ward's® Granite (Gray)
Catalog # 470026-012
Supplier:  Avantor
Ward's® Granite (Gray)
Ward's® Granite (Gray)
Granite (Gray), Bulk Pack, 1 kg, Size will vary
Price
$313.85
Log in for availability
Each (1UN)
Product Details & Documents
Fine to medium-grained with biotite.
  • Collected in Vermont
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student Specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk Pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Specifications
  • Description:
    Granite (Gray), Bulk Pack, 1 kg
  • Specimen Dimensions:
    Size will vary