Ward's® Igneous Rock Collection
Igneous Rock Collection, 12
Catalog # 470025-222
Supplier: Avantor
Jump to:
Product Details & Documents
Samples of Intrusive and Extrusive Igneous Rocks
- Introduce concepts of the Earth's crust
- Compartmented box
Provide your students with an excellent introduction to the forces at work beneath the Earth’s crust. The set is housed in a compartmented collection box and includes an identification list.
Contents:
12 numbered samples, 1" x 2"
Rocks:
basalt, diorite, gabbro, granite, obsidian, pegmatite, peridotite, pumice, rhyolite (2), scoria, syenite
Hand Specimen Set:
Large hand samples are ideal for demonstration and display. This set is housed in individual, labeled plastic display trays and comes with an identification list.
Contents:
12 numbered samples, 3" x 4"
Documents
Igneous Rock Collection
Additional Product Details
Specifications
- Description:Igneous Rock Collection
- No. of Samples:12