Order Entry
Mexico
Orders LinkContactUsLinkComponent
Ward's® Scoria
Ward's® Scoria
Scoria thin section
Catalog # 470025-032
Supplier:   Avantor
Ward's® Scoria
Catalog # 470025-032
Supplier:  Avantor
Ward's® Scoria
Ward's® Scoria
Scoria thin section
Price
$917.00
Log in for availability
Each (1UN)
Product Details & Documents
Gray-to-brown basaltic, with over 50% vesicles.
  • Collected in Oregon
  • Multiple sizes and pack options to fit specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student Specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk Pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" - 2×3". Packaged in a labeled storage box for easy organization.
All Specimen dimensions are considered an average size range.
Specifications
  • Description:
    Scoria thin section