Order Entry
Mexico
Orders LinkContactUsLinkComponent
Ward's® Apatite (Massive)
Apatite thin section, —
  470024-886
 :   Avantor
Ward's® Apatite (Massive)
  470024-886
 :  Avantor
Ward's® Apatite (Massive)
Ward's® Apatite (Massive)
Apatite thin section, —
$1,043.40
Each (1UN)
Blue fluorapatite, pure crystalline, Moh's hardness of 5.
  • Collected in Madagascar
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25 to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
  • Description:
    Apatite thin section
  • Specimen Dimensions:
  • Cat. No.:
    470024-886