Ward's® Killing Jars
WARD’S Killing Jars, 16oz. Jar
Catalog # 470018-466
Jump to:
Discontinued
Product Details & Documents
The improved design of these jars incorporates a bottom pad that better absorbs killing agent, so that fumes are contained and insects are knocked down quickly, causing less damage to specimens.
- Absorbent pad contains the fumes
Just one application lasts for hours, and the permanent label provides instructions for use.
Specifications
- Description:WARD’S Killing Jars, 16oz. Jar