Ward's® Coal (Peat)
Coal (Peat), Hand Specimens, 2×3" - 3×4"
Catalog # 470015-486
Supplier: Avantor
Jump to:
Product Details & Documents
Compact Plant Fiber Masses.
Ordering information: WARD’S offers different specimen sizes to meet your various needs. Hand specimens are great for lectures, demonstrations, and display. Hand specimens range in size from 2×3" to 3×4", and each comes in a labeled plastic tray. Bulk pack includes several of the smaller pieces that are suitable for classroom study, while the larger specimens in the one kilogram pack can be cut to sizes that meet your specific needs. The number and size of pieces will vary.
Specifications
- Description:Coal (Peat), Hand Specimens
- Specimen Dimensions:2×3" - 3×4"
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



