Ward's® Coal (Lignite)
Coal (Lignite), student specimens, 1×1" - 1×1"
470015-464
: Avantor
Brownish black, intermediate stage between peat and bituminous coal.
- Collected in North Dakota
- Multiple sizes and pack options to fit your specific needs
- Cut by Ward's geologists to guarantee top market quality
Student Specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk Pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
- Description:Coal (Lignite), student specimens
- Specimen Dimensions:1×1" - 1×1"